Ammoniumsulfat, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Synonyms: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite
1.01216.5000 1.01216.9029 1.01216.9027
1.01216.5000EA 7425 SEK
1.01216.5000 1.01216.9029 1.01216.9027
Ammoniumsulfat, SAFC®
Ammonium sulphate
Formel: (NH₄)₂SO₄
Smältpunkt: 336…339 °C
Densitet: 1,78 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003391
CAS nummer: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559