Ammoniumsulfat ≥99.999%, Puratronic®
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite
10639.09 10639.18 10639.30
10639.09EA 710 SEK
10639.09 10639.18 10639.30
Ammoniumsulfat ≥99.999%, Puratronic®
Ammonium sulphate
Purity: 99.999% CAS No.: 7783-20-2 Molecular Formula: H8N2O4S Formula Weight: 132.14 Physical Form: Crystalline Density: 1.769 Appearance: White Melting Point: ca 280° dec. Merck Reference: 14,555 MDL No.: MFCD00003391
Formel: (NH₄)₂SO₄
MW: 132.14 g/mol
Smältpunkt: ∼ 280 °C (dec.)
Densitet: 1.769
Storage Temperature: Omgivande
MDL Number: MFCD00003391
CAS nummer: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 14,555