Ammoniumtiocyanat ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Thiocyanic acid ammonium salt, Ammonium rhodanide, Ammonium sulphocyanate
A10632.0B A10632.0I A10632.30
A10632.0BEA 1575 SEK
A10632.0B A10632.0I A10632.30
Ammoniumtiocyanat ≥98%
Ammoniumtiocyanat
Purity: 98+% CAS No.: 1762-95-4 Molecular Formula: CH4N2S Formula Weight: 76.12 Odour: Odourless Melting Point: 147-149° Beilstein Reference: 3595135 Merck Reference: 14,561 MDL No.: MFCD00011428
Formel: NH₄SCN
MW: 76.12 g/mol
Smältpunkt: 147…149 °C
Densitet: 1.305
Storage Temperature: Omgivande
MDL Number: MFCD00011428
CAS nummer: 1762-95-4
EINECS: 217-175-6
Merck Index: 13,00564