Diammoniumperoxodisulfat (APS)
Leverantör: Molekula
|
Fara
|
19148668-1KG
19148668-100G
MOLE19148668-1KGEA
0
SEK
MOLE19148668-1KG
MOLE19148668-100G
Diammoniumperoxodisulfat (APS)
Diammoniumperoxodisulfat (APS)
Formel:
(NH₄)₂S₂O₈ Smältpunkt: 120 °C Densitet: 1,98 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00003390 CAS nummer: 7727-54-0 EINECS: 231-786-5 UN: 1444 ADR: 5.1,III Merck Index: 13,00545 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...