Ammoniumkarbonat (∼30% NH₃ )
Leverantör: Thermo Fisher Scientific
Varning
A13955.0B A13955.0I
A13955.0BEA 870 SEK
A13955.0B A13955.0I
Ammoniumkarbonat (∼30% NH₃ )
Ammoniumkarbonat
CAS No.: 506-87-6 Molecular Formula: CH8N2O3 Formula Weight: 96.09 Odour: Ammonia-like Melting Point: 60° subl. Beilstein Reference: 3627235 Merck Reference: 14,508 MDL No.: MFCD00010890
Formel: (NH₄)₂CO₃
Smältpunkt: 60 °C (subl.)
Densitet: 1,5 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010890
CAS nummer: 506-87-6
EINECS: 208-058-0
Merck Index: 14,508