Ammoniumklorid, EMPROVE® EXPERT BP, Ph. Eur., USP suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Varning
Synonyms: Ammonium muriate, Salmiac
1.01142.5000 1.01142.9025
1.01142.5000EA 5450 SEK
1.01142.5000 1.01142.9025
Ammoniumklorid, EMPROVE® EXPERT BP, Ph. Eur., USP suitable for use as excipient, SAFC®
Ammoniumklorid
Formel: NH₄Cl
MW: 53.49 g/mol
Smältpunkt: 335
Densitet: 1.5256 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011420
CAS nummer: 12125-02-9
EINECS: 235-186-4
Merck Index: 13,00510
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM