Tetraphenylphosphonium iodide ≥98%
Leverantör: Thermo Fisher Scientific
Varning
A11539.06 A11539.14 A11539.22
A11539.06EA 1325 SEK
A11539.06 A11539.14 A11539.22
Tetraphenylphosphonium iodide ≥98%
Tetraphenylphosphonium iodide
Purity: 98+% CAS No.: 2065-67-0 Molecular Formula: (C6H5)4PI Formula Weight: 466.31 Melting Point: 345-350° Beilstein Reference: 3922216
Formel: C₂₄H₂₀IP
Smältpunkt: 345…350 °C
Densitet: 1,5 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011917
CAS nummer: 2065-67-0
EINECS: 218-179-0