Triphenylfosfin 99%
Leverantör: Thermo Fisher Scientific
Varning
140420010 140420250 140422500 140420050
ACRO140420010EA 3500 SEK
ACRO140420010 ACRO140420250 ACRO140422500 ACRO140420050
Triphenylfosfin 99%
Triphenylfosfin
Formel: C₁₈H₁₅P
Kokpunkt: 377 °C (760 mmHg)
Densitet: 1,194 g/cm³ (20 °C)
Flampunkt: 181 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003043
CAS nummer: 603-35-0
EINECS: 210-036-0
Merck Index: 12,09873
Order Now
Specifikation testresultat
Appearance White Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 78.5 to 81.5 °C
GC ≥97.5 %