Triphenylarsine zS, Sigma-Aldrich®
Leverantör: Merck
|
Fara
|
MQ200
8.08653.0025
8.08653.0005
8.08653.0025EA
2100
SEK
8.08653.0025
8.08653.0005
Triphenylarsine zS, Sigma-Aldrich®
Triphenylarsine
Formel:
C₁₈H₁₅As MW: 306.24 g/mol Kokpunkt: ∼360 °C (1013 hPa) Smältpunkt: 58 Densitet: 1.306 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00002994 CAS nummer: 603-32-7 EINECS: 210-032-9 UN: 3465 ADR: 6.1 II |
Specifikation testresultat
Solubility | (20 °C) insoluble |
Melting Point | 59 - 61 °C |
Molar Mass | 306.24 g/mol |
Boiling Point |
|
Flash Point | 265 °C |
Density | 1.22 g/cm3 (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...