Tris(3-methylphenyl)phosphine ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Tris(m-tolyl)phosphine, Tri-m-tolylphosphine
A15372.03 A15372.06 A15372.14
A15372.03EA 915 SEK
A15372.03 A15372.06 A15372.14
Tris(3-methylphenyl)phosphine ≥98%
Tris(3-methylphenyl)phosphine
Purity: 98+% CAS No.: 6224-63-1 Molecular Formula: C21H21P Formula Weight: 304.37 Melting Point: 99-101° Beilstein Reference: 658863
Formel: C₂₁H₂₁P
MW: 304,37 g/mol
Smältpunkt: 99…101 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008532
CAS nummer: 6224-63-1
EINECS: 228-312-4