Tris(2-methylphenyl)phosphine ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Tris(o-tolyl)phosphine, Tri-o-tolylphosphine
A12093.06 A12093.03 A12093.14
A12093.06EA 2875 SEK
A12093.06 A12093.03 A12093.14
Tris(2-methylphenyl)phosphine ≥98%
Tris(2-methylphenyl)phosphine
Purity: 98+% CAS No.: 6163-58-2 Molecular Formula: C21H21P Formula Weight: 304.37 Melting Point: 122-127° Beilstein Reference: 661212
Formel: C₂₁H₂₁P
MW: 304,37 g/mol
Kokpunkt: 187…192 °C (0,05 mmHg)
Smältpunkt: 122…127 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008514
CAS nummer: 6163-58-2
EINECS: 228-193-9