Chlorodiphenylphosphine 98%

Leverantör: Thermo Fisher Scientific
Fara

Synonyms: DPPC, Diphenylchlorophosphine

347300050 347300250 347301000 347305000
ACRO347300050EA 420 SEK
ACRO347300050 ACRO347300250 ACRO347301000 ACRO347305000
Chlorodiphenylphosphine 98%
Chlorodiphenylphosphine
Formel: C₁₂H₁₀ClP
Kokpunkt: 320 °C (760 mmHg)
Smältpunkt: 14 - 16 °C
Densitet: 1,229 g/cm³
Flampunkt: 138 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000529
CAS nummer: 1079-66-9
EINECS: 214-093-2
UN: 3265
ADR: 8,II

Order Now

Specifikation testresultat

Appearance Colorless to yellow Liquid (May develop some turbidity or precipitate)
Infrared spectrum Conforms
Refractive index 1.6350 to 1.6370 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR