Benzyltriethylammonium bromide ≥98%
Leverantör: Thermo Fisher Scientific
Varning
A14291.14 A14291.22 A14291.36
A14291.14EA 700 SEK
A14291.14 A14291.22 A14291.36
Benzyltriethylammonium bromide ≥98%
Benzyltriethylammonium bromide
Purity: 98+% CAS No.: 5197-95-5 Molecular Formula: (CH3CH2)3N(CH2C6H5)Br Formula Weight: 272.24 Melting Point: ca 200° dec. Beilstein Reference: 3739113
Formel: C₁₃H₂₂BrN
MW: 272.23 g/mol
Smältpunkt: ∼ 200 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00011822
CAS nummer: 5197-95-5
EINECS: 225-986-1