Benzyltriethylammonium bromide 99%
Leverantör: Thermo Fisher Scientific
Varning
164200250 164201000
ACRO164200250EA 645 SEK
ACRO164200250 ACRO164201000
Benzyltriethylammonium bromide 99%
Benzyltriethylammonium bromide
Formel: C₁₃H₂₂BrN
MW: 272.23 g/mol
Smältpunkt: 187…197 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011822
CAS nummer: 5197-95-5
EINECS: 225-986-1
Order Now
Specifikation testresultat
Appearance White to cream to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 187 to 197 °C (Dec)
Titration Argentometric ≥98.5 %