Tetrabutylammonium fluoride trihydrate ≥98%
Leverantör: Thermo Fisher Scientific
Fara
L13303.09 L13303.18
L13303.09EA 1025 SEK
L13303.09 L13303.18
Tetrabutylammonium fluoride trihydrate ≥98%
Tetrabutylammonium fluoride trihydrate
Purity: 98% CAS No.: 87749-50-6 Molecular Formula: C16H36FN.3H2O Formula Weight: 315.52 (261.47anhy) Odour: Odourless Melting Point: 54-60° Beilstein Reference: 3761900 UN No.: 2811 MDL No.: MFCD00149981
Formel: C₁₆H₃₆FN·3H₂O
MW: 315.51 g/mol
Smältpunkt: 54…60 °C
Storage Temperature: Omgivande
MDL Number: MFCD00149981
CAS nummer: 87749-50-6
EINECS: 207-057-2
UN: 1759
ADR: 8 III