Tetrametylammoniumklorid 98+%
Leverantör: Thermo Scientific
Fara
Synonyms: TMACl, TMA
138331000 138332500 138330010
ACRO138331000EA 605 SEK
ACRO138331000 ACRO138332500 ACRO138330010
Tetrametylammoniumklorid 98+%
Tetrametylammoniumklorid
Formel: (CH₃)₄NCl
MW: 109,60 g/mol
Smältpunkt: 420 °C
Densitet: 1,169 g/cm³ (20 °C)
Flampunkt: ≥ 100 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011628
CAS nummer: 75-57-0
EINECS: 200-880-8
UN: 2811
ADR: 6.1,II
Order Now
Specifikation testresultat
Appearance White to cream Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥98.0 %
Stabilizer ≤0.05 % tert-Butylcatechol