Tetrabutylammonium chloride xhydrate
Leverantör: Thermo Scientific
Varning
Synonyms: Tetrabutyl ammonium chloride xhydrate, Tetra-n-butylammonium chloride hydrate, Tetrabutylammonium chloride hydrate, TBAC
19347.06 19347.14 19347.22
19347.06EA 615 SEK
19347.06 19347.14 19347.22
Tetrabutylammonium chloride xhydrate
Tetrabutylammonium chloride xhydrate
CAS No.: 37451-68-6 Molecular Formula: [CH3(CH2)3]4NCl.xH2O Formula Weight: 277.92(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Melting Point: 73-75° MDL No.: MFCD00011635
Formel: C₁₆H₃₆ClN·xH₂O
MW: 277,92 g/mol (anhy)
Smältpunkt: 73…75 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011635
CAS nummer: 37451-68-6
EINECS: 214-195-7