Tetrametylammoniumbromid ≥98%
Leverantör: Thermo Scientific
Fara
A13368.22 A13368.0E A13368.36
A13368.22EA 740 SEK
A13368.22 A13368.0E A13368.36
Tetrametylammoniumbromid ≥98%
Tetrametylammoniumbromid
Purity: 98% CAS No.: 64-20-0 Molecular Formula: C4H12BrN Formula Weight: 154.06 Odour: Odourless Melting Point: 300° Beilstein Reference: 3620955 UN No.: 2811 MDL No.: MFCD00011626
Formel: C₄H₁₂BrN
MW: 154,05 g/mol
Kokpunkt: ∼360 °C (1013 hPa)
Smältpunkt: >300 °C
Densitet: 1,56
Storage Temperature: Omgivande
MDL Number: MFCD00011626
CAS nummer: 64-20-0
EINECS: 200-581-2
UN: 2811
ADR: 6.1,II