Tetramethylammonium tetrafluoroborate ≥97%

Leverantör: Thermo Scientific
Varning

Synonyms: Tetramethylammoniumtetrafluoroborate

A17436.14 A17436.06 A17436.22
A17436.14EA 2000 SEK
A17436.14 A17436.06 A17436.22
Tetramethylammonium tetrafluoroborate ≥97%
Tetramethylammonium tetrafluoroborate

Purity: 97% CAS No.: 661-36-9 Molecular Formula: (CH3)4NBF4 Formula Weight: 160.95 Melting Point: 417-419° Beilstein Reference: 3715404

Formel: C₄H₁₂BF₄N
MW: 160,95 g/mol
Smältpunkt: 417…419 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011745
CAS nummer: 661-36-9
EINECS: 211-547-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR