Tetrabutylammonium tetrafluoroborate 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Tetrabutylammoniumtetrafluoroborate
353090250 353091000 353090050
ACRO353090250EA 1425 SEK
ACRO353090250 ACRO353091000 ACRO353090050
Tetrabutylammonium tetrafluoroborate 98%
Tetrabutylammonium tetrafluoroborate
Formel: C₁₆H₃₆BF₄N
MW: 329.27 g/mol
Smältpunkt: 156…163 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011634
CAS nummer: 429-42-5
EINECS: 207-058-8
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Infrared spectrum Conforms
Assay ≥97.5 %