Tributylphosphine 95%
Leverantör: Thermo Scientific
Fara
Synonyms: Tri-n-butylphosphine, TBP
139341000 139345000
ACRO139341000EA 1400 SEK
ACRO139341000 ACRO139345000
Tributylphosphine 95%
Tributylphosphine
Formel: C₁₂H₂₇P
MW: 202,32 g/mol
Kokpunkt: 240…242 °C (760 mmHg)
Smältpunkt: –65 °C
Densitet: 0,81 g/cm³
Flampunkt: 37 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009462
CAS nummer: 998-40-3
EINECS: 213-651-2
UN: 3254
ADR: 4.2,I
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4610 to 1.4630 (20°C, 589 nm)
GC ≥94 %