Trioctylfosfinoxid 99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: TOPO, Tri-n-octylphosphine Oxide, Trioctylphosphinoxide
207600250 207601000 207605000
ACRO207600250EA 1075 SEK
ACRO207600250 ACRO207601000 ACRO207605000
Trioctylfosfinoxid 99%
Trioctylfosfinoxid
Formel: C₂₄H₅₁OP
MW: 386.64 g/mol
Kokpunkt: 201…202 °C (2 mmHg)
Smältpunkt: 50…55 °C
Flampunkt: 230 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002083
CAS nummer: 78-50-2
EINECS: 201-121-3
Order Now
Specifikation testresultat
Appearance White to light yellow Powder or crystals or flakes and/or chunks
Infrared spectrum Conforms
Melting point 50 to 55 °C
GC ≥98.5 % (including isomer)
Isomer ≤6 %