(Methyl)triphenylphosphonium bromide ≥98%
Leverantör: Thermo Scientific
Varning
A15878.30 A15878.0B A15878.18
A15878.30EA 2425 SEK
A15878.30 A15878.0B A15878.18
(Methyl)triphenylphosphonium bromide ≥98%
(Methyl)triphenylphosphonium bromide
Purity: 98+% CAS No.: 1779-49-3 Molecular Formula: CH3P(C6H5)3Br Formula Weight: 357.24 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 231-234° Beilstein Reference: 3599467 UN No.: 2811 MDL No.: MFCD00011804
Formel: C₁₉H₁₈BrP
MW: 357,23 g/mol
Smältpunkt: 231…234 °C
Flampunkt: >240 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011804
CAS nummer: 1779-49-3
EINECS: 217-218-9
UN: 2811
ADR: 6.1,III