Antimon(III)sulfid ≥99.9%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Needle antimony, Antimony(III) sulphide, Amtimony sulphide, Diantimony trisulphide, Stibnite, Antimony glance, Antimony trisulphide, Antimonous sulphide, Antimony sulphide black
40371.09 40371.18 40371.30
40371.09EA 1950 SEK
40371.09 40371.18 40371.30
Antimon(III)sulfid ≥99.9%
Antimony (III) sulphide
Purity: 99.9% CAS No.: 1345-04-6 Molecular Formula: Sb2S3 Formula Weight: 339.68 Physical Form: 3-12mm Pieces Melting Point: 550° UN No.: 1549 MDL No.: MFCD00011217
Vacuum deposition
Formel: S₃Sb₂
MW: 339.72 g/mol
Kokpunkt: 1150
Smältpunkt: 550
Densitet: 4.12 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011217
CAS nummer: 1345-04-6
EINECS: 215-713-4
UN: 1549
ADR: 6.1 III
Merck Index: 14,713