Triisopropyl phosphite 96%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Tri-i-propylphosphite
372381000 372385000 372380025
ACRO372381000EA 1025 SEK
ACRO372381000 ACRO372385000 ACRO372380025
Triisopropyl phosphite 96%
Triisopropyl phosphite
Formel: C₉H₂₁O₃P
MW: 208.24 g/mol
Kokpunkt: 94…95 °C (66.6hPa)
Densitet: 0.914 g/cm³
Flampunkt: 62 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008874
CAS nummer: 116-17-6
EINECS: 204-130-0
UN: 3278
ADR: 6.1 III
Order Now
Specifikation testresultat
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4095 to 1.4115 (20°C, 589 nm)
GC ≥95 %
Color scale ≤25 APHA