Trietylfosfat
Leverantör: EHRENSTORFER
|
Varning
|
C17835500
EHERC17835500EA
930
SEK
EHERC17835500
Trietylfosfat
Trietylfosfat
Formel:
C₆H₁₅O₄P MW: 182.16 g/mol Kokpunkt: 215 Smältpunkt: –57 °C Densitet: 1.07 g/cm³ (20 °C) Flampunkt: 116 Storage Temperature: Omgivande |
MDL Number:
MFCD00009077 CAS nummer: 78-40-0 EINECS: 201-114-5 Merck Index: 12,09806 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...