Diethyl phosphonate 98%
Leverantör: Thermo Fisher Scientific
Fara
114500010 114502500 114500025 114500000
ACRO114500010EA 2050 SEK
ACRO114500010 ACRO114502500 ACRO114500025 114500000.
Diethyl phosphonate 98%
Diethyl phosphonate
Formel: C₄H₁₁O₃P
MW: 138.1 g/mol
Kokpunkt: 60 °C (6mbar)
Smältpunkt: –70 °C
Densitet: 1.08 g/cm³
Flampunkt: 74 °C
Storage Temperature: Omgivande
MDL Number: MFCD00044573
CAS nummer: 762-04-9
EINECS: 212-091-6
Order Now
Specifikation testresultat
Appearance Colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4070 to 1.4090 (20°C, 589 nm)
GC ≥98.5 %