Antimony (V) oxide ≥99.998%, Puratronic®
Leverantör: Thermo Scientific
Varning
Synonyms: Antimony pentoxide, Diantimony pentoxide
10643.06 10643.14 10643.22
10643.06EA 1125 SEK
10643.06 10643.14 10643.22
Antimony (V) oxide ≥99.998%, Puratronic®
Antimony (V) oxide
Purity: 99.998% CAS No.: 1314-60-9 Molecular Formula: Sb2O5 Formula Weight: 323.50 Physical Form: Powder Density: 3.78 Melting Point: 380° dec. Solubility: Slightly soluble in water. Slowly dissolves in warm HCl and warm KOH. Insoluble in HNO3 Merck Reference: 14,698 UN No.: 1549 MDL No.: MFCD00011216
Formel: O₅Sb₂
MW: 323,52 g/mol
Smältpunkt: 380 °C (dec.)
Densitet: 3,78
Storage Temperature: Omgivande
MDL Number: MFCD00011216
CAS nummer: 1314-60-9
EINECS: 215-237-7
UN: 1549
ADR: 6.1,III
Merck Index: 14,698