Vismut(III)nitrat pentahydrat 98
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Bismuth trinitrate pentahydrate
A11748.22 A11748.36
A11748.22EA 775 SEK
A11748.22 A11748.36
Vismut(III)nitrat pentahydrat 98
Vismut(III)nitrat pentahydrat
Purity: 98% CAS No.: 10035-06-0 Molecular Formula: Bi(NO3)3.5H2O Formula Weight: 485.07 (394.99anhy) Odour: Odourless Melting Point: 30° dec. UN No.: 1477 MDL No.: MFCD00149157
Formel: Bi(NO₃)₃·5H₂O
MW: 485.07 g/mol
Kokpunkt: 75…80 °C (1013 hPa)
Smältpunkt: 30 °C (dec.)
Densitet: 2.83
Storage Temperature: Omgivande
MDL Number: MFCD00149157
CAS nummer: 10035-06-0
EINECS: 233-791-8
UN: 1477
ADR: 5.1 II