Vismut(III)nitrat pentahydrat ≥98% ACS
Leverantör: Thermo Scientific
Fara
Synonyms: Bismuth trinitrate pentahydrate
12231.30 12231.A1
12231.30EA 1575 SEK
12231.30 12231.A1
Vismut(III)nitrat pentahydrat ≥98% ACS
Vismut(III)nitrat pentahydrat
Application in luminous paints, preparation of bismuth salts
Purity: 98% CAS No.: 10035-06-0 Molecular Formula: Bi(NO3)3.5H2O Formula Weight: 485.07 (394.99anhy) Physical Form: Crystalline Density: 2.83 Melting Point: 30° dec. Solubility: Soluble in dilute HNO3 acid solution. Decomposes in water to subnitrate. Soluble in dilute acetic acid, glycerol, acetone. Insoluble in alcohol, ethyl acetate Merck Reference: 14,1271 UN No.: 1477 MDL No.: MFCD00149157
Luminous paints, preparation of bismuth salts
Formel: Bi(NO₃)₃·5H₂O
MW: 485,07 g/mol
Kokpunkt: 75…80 °C (1013 hPa)
Smältpunkt: 30 °C (dec.)
Densitet: 2,83
Storage Temperature: Omgivande
MDL Number: MFCD00149157
CAS nummer: 10035-06-0
EINECS: 233-791-8
UN: 1477
ADR: 5.1,II
Merck Index: 14,1271