Bismuth(III) fluoride, vattenfri 99%
Leverantör: Thermo Fisher Scientific
Fara
13075.14 13075.22 13075.36
13075.14EA 1250 SEK
13075.14 13075.22 13075.36
Bismuth(III) fluoride, vattenfri 99%
Bismuth(III) fluoride
Purity: 99% CAS No.: 7787-61-3 Molecular Formula: BiF3 Formula Weight: 265.98 Physical Form: Powder Refractive Index: 1.74 Sensitivity: Hygroscopic Odour: Odourless Melting Point: 727° Solubility: Practically insoluble in water. Soluble in HF Merck Reference: 14,1265 UN No.: 3260 MDL No.: MFCD00010893
Manufacturing of bismuth pentafluoride
Formel: BiF₃
MW: 265.98 g/mol
Kokpunkt: 230
Smältpunkt: 727 °C
Densitet: 3.7 g/cm³ (160 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010893
CAS nummer: 7787-61-3
EINECS: 232-124-8
UN: 3260
ADR: 8 III
Merck Index: 14,1265