Antimontrifluorid 98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Antimony trifluoride
191241000 191245000
ACRO191241000EA 900 SEK
ACRO191241000 ACRO191245000
Antimontrifluorid 98%
Antimontrifluorid
Formel: F₃Sb
MW: 178.76 g/mol
Kokpunkt: 376
Smältpunkt: 292
Storage Temperature: Omgivande
MDL Number: MFCD00011218
CAS nummer: 7783-56-4
EINECS: 232-009-2
UN: 2923
ADR: 8 II
Merck Index: 13,00715
Order Now
Specifikation testresultat
Appearance White to grey Crystalline powder
Titration Iodimetric ≥97.5 %