Bismuth(III) bromide ≥99%
Leverantör: Thermo Scientific
Fara
17112.14 17112.22
17112.14EA 2925 SEK
17112.14 17112.22
Bismuth(III) bromide ≥99%
Bismuth(III) bromide
Purity: 99% CAS No.: 7787-58-8 Molecular Formula: BiBr3 Formula Weight: 448.71 Physical Form: Powder Density: 5.72 Appearance: Yellow Odour: Acidic Melting Point: 218° Boiling Point: 453° UN No.: 3260 MDL No.: MFCD00014195
Formel: BiBr₃
MW: 448,69 g/mol
Kokpunkt: 453 °C
Smältpunkt: 218 °C
Densitet: 5,72
Storage Temperature: Omgivande
MDL Number: MFCD00014195
CAS nummer: 7787-58-8
EINECS: 232-121-1
UN: 3260
ADR: 8,III
Merck Index: 12,01299