Tetrametylsilan 99.9%
Leverantör: Thermo Scientific
Fara
Synonyms: TMS
A13148.14 A13148.22
A13148.14EA 945 SEK
A13148.14 A13148.22
Tetrametylsilan 99.9%
Tetrametylsilan
Purity: 99.9% CAS No.: 75-76-3 Molecular Formula: C4H12Si Formula Weight: 88.23 Storage Temperature: 2°C to 8°C Flash Point: -27°(-16°F) Refractive Index: 1.3590 Odour: Mild Melting Point: -100° Boiling Point: 26-27° Beilstein Reference: 1696908 Merck Reference: 14,9229 UN No.: 2749 MDL No.: MFCD00008274
Formel: C₄H₁₂Si
MW: 88,22 g/mol
Kokpunkt: 26…27 °C
Smältpunkt: –100 °C
Densitet: 0,646
Flampunkt: –27 °C (–16 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00008274
CAS nummer: 75-76-3
EINECS: 200-899-1
UN: 2749
ADR: 3,I
Merck Index: 14,9229