Tetrametylsilan 99.9+% for NMR spectroscopy
Leverantör: Thermo Scientific
Fara
Synonyms: TMS
138470250 138471000
ACRO138470250EA 820 SEK
ACRO138470250 ACRO138471000
Tetrametylsilan 99.9+% for NMR spectroscopy
Tetrametylsilan
Formel: C₄H₁₂Si
MW: 88,22 g/mol
Kokpunkt: 26…28 °C (760 mmHg)
Smältpunkt: –99 °C
Densitet: 0,64 g/cm³
Flampunkt: –27 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00008274
CAS nummer: 75-76-3
EINECS: 200-899-1
UN: 2749
ADR: 3,I
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
GC ≥99.90 %
Suitability (for 1H-NMR position ref.) Passes test
Elemental Analysis 6.09 to 6.59 % N