Klortrimetylsilan 98
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: TMCS, TMSC, Trimethylchlorosilane
A13651.AC A13651.AE A13651.AP A13651.AU
A13651.ACEA 550 SEK
A13651.AC A13651.AE A13651.AP A13651.AU
Klortrimetylsilan 98
Klortrimetylsilan
Purity: 98+% CAS No.: 75-77-4 Molecular Formula: C3H9ClSi Formula Weight: 108.64 Flash Point: -27°(-16°F) Refractive Index: 1.3887 Odour: Chlorine-like Melting Point: -58° Boiling Point: 56-58° Beilstein Reference: 1209232 UN No.: 1298 MDL No.: MFCD00000502
Formel: C₃H₉ClSi
MW: 108.64 g/mol
Kokpunkt: 56-58 °C
Smältpunkt: –58 °C
Densitet: 0.856
Flampunkt: –27 °C (–16 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000502
CAS nummer: 75-77-4
EINECS: 200-900-5
UN: 1298
REACH: 01-2119457596-25