Tributyltin chloride 95% tech.
Leverantör: Thermo Scientific
Fara
Synonyms: Chlorotributylstannane, Tri-n-butyltin chloride, TBTC
139351000 139355000
ACRO139351000EA 835 SEK
ACRO139351000 ACRO139355000
Tributyltin chloride 95% tech.
Tributyltin chloride
Formel: C₁₂H₂₇ClSn
MW: 325,51 g/mol
Kokpunkt: 171…173 °C (25 mmHg)
Smältpunkt: –9 °C
Densitet: 1,2 g/cm³
Flampunkt: >112 °C
MDL Number: MFCD00000521
CAS nummer: 1461-22-9
EINECS: 215-958-7
UN: 2788
ADR: 6.1,II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4880 to 1.4940 (20°C, 589 nm)
Titration Argentometric ≥94 %