Diklordioktylstannan ≥95%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Dichlorodioctylstannane, Di-n-octyldichlorotin
41738.06 41738.14 41738.22
41738.06EA 1475 SEK
41738.06 41738.14 41738.22
Diklordioktylstannan ≥95%
Diklordioktylstannan
Purity: 95% CAS No.: 3542-36-7 Molecular Formula: [CH3(CH2)7]2SnCl2 Formula Weight: 416.04 Physical Form: Crystalline Sensitivity: Moisture Sensitive Boiling Point: 175°/1mm UN No.: 3261 MDL No.: MFCD00018091
Formel: C₁₆H₃₄Cl₂Sn
MW: 416.06 g/mol
Kokpunkt: 175 °C (1 mmHg)
Smältpunkt: 44…48 °C
Storage Temperature: Omgivande
MDL Number: MFCD00018091
CAS nummer: 3542-36-7
EINECS: 222-583-2
UN: 3261