Chlorodimethylsilane 97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Dimethylchlorosilane
162840100 162840500 162842500
ACRO162840100EA 385 SEK
ACRO162840100 ACRO162840500 ACRO162842500
Chlorodimethylsilane 97%
Chlorodimethylsilane
Formel: C₂H₇ClSi
MW: 94.62 g/mol
Kokpunkt: 34.6 °C (760 mmHg)
Smältpunkt: –111 °C
Densitet: 0.85 g/cm³
Flampunkt: –28 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000495
CAS nummer: 1066-35-9
EINECS: 213-912-0
UN: 2924
ADR: 3 I
REACH: 01-2119486782-25
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3815 to 1.3855 (20°C, 589 nm)
GC ≥96.0 %