Methyltrimethoxysilane 97%
Leverantör: Thermo Scientific
Fara
Synonyms: Trimethoxy(methyl)silane
250540050 250542500 250540010
ACRO250540050EA 415 SEK
ACRO250540050 ACRO250542500 ACRO250540010
Methyltrimethoxysilane 97%
Methyltrimethoxysilane
Formel: C₄H₁₂O₃Si
MW: 136,22 g/mol
Kokpunkt: 102 °C (760 mmHg)
Smältpunkt: –50 °C
Densitet: 0,95 g/cm³
Flampunkt: 11 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008342
CAS nummer: 1185-55-3
EINECS: 214-685-0
UN: 1993
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3695 to 1.3715 (20°C, 589 nm)
GC ≥96.0 %