Dodecyltriethoxysilane 95%

Leverantör: Thermo Fisher Scientific
Varning

L17293.22 L17293.14
L17293.22EA 7125 SEK
L17293.22 L17293.14
Dodecyltriethoxysilane 95%
Dodecyltriethoxysilane

Purity: 95% CAS No.: 18536-91-9 Molecular Formula: C18H40O3Si Formula Weight: 332.60 Flash Point: 110°(230°F) Refractive Index: 1.4330 Boiling Point: 152-153°/3mm Beilstein Reference: 2414223 MDL No.: MFCD00048437

Formel: C₁₈H₄₀O₃Si
Kokpunkt: 152…153 °C (3 mmHg)
Densitet: 0,890
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00048437
CAS nummer: 18536-91-9
EINECS: 242-409-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR