Isobutyltrimethoxysilane ≥97%
Leverantör: Thermo Fisher Scientific
Varning
B21048.14 B21048.22 B21048.36
B21048.14EA 565 SEK
B21048.14 B21048.22 B21048.36
Isobutyltrimethoxysilane ≥97%
Isobutyltrimethoxysilane
Purity: 97% CAS No.: 18395-30-7 Molecular Formula: (CH3)2CHCH2Si(OCH3)3 Formula Weight: 178.31 Flash Point: 43°(109°F) Refractive Index: 1.3960 Boiling Point: 154-157° Beilstein Reference: 2233976 UN No.: 1993 MDL No.: MFCD00051589
Formel: (H₃C)₂HCSi(OCH₃)₃
Kokpunkt: 154…157 °C
Densitet: 0,930
Flampunkt: 43 °C (109 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00051589
CAS nummer: 18395-30-7
EINECS: 242-272-5
ADR: 3,III