(3-Chloropropyl)trimethoxysilane ≥97%
Leverantör: Thermo Fisher Scientific
|
Varning
|
Purity: 97%
CAS No.: 2530-87-2
Molecular Formula: ClCH2CH2CH2Si(OCH3)3
Formula Weight: 198.72
Flash Point: 78°(172°F)
Refractive Index: 1.4200
Odour: Recognizable
Melting Point: -50°
Boiling Point: 195-197°
Beilstein Reference: 969627
MDL No.: MFCD00000997
Formel:
ClH₂CCH₂CH₂Si(OCH₃)₃ Kokpunkt: 195…197 °C Smältpunkt: –50 °C Densitet: 1,077 Flampunkt: 78 °C (172 °F) Storage Temperature: Omgivande |
MDL Number:
MFCD00000997 CAS nummer: 2530-87-2 EINECS: 219-787-9 |