Silicon dioxide (Silica, vitreous), granulat, Patinal® 2.5-4 mm

Leverantör: Merck

Synonyms: Quartz, Sand, Cristobalite

1.15337.0100 1.15337.1000
1.15337.0100EA 0 SEK
1.15337.0100 1.15337.1000
Silicon dioxide (Silica, vitreous), granulat, Patinal® 2.5-4 mm
Silicon dioxide (Silica, vitreous)
Formel: SiO₂
MW: 60,08 g/mol
Kokpunkt: 2230 °C (1013 hPa)
Smältpunkt: 1710 °C
Densitet: 2,196 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011232
CAS nummer: 60676-86-0
EINECS: 262-373-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR