Kiselgel ≥99.0%
Leverantör: Thermo Scientific
Varning
Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
89709.A1 89709.A7 89709.30
89709.A1EA 5650 SEK
89709.A1 89709.A7 89709.30
Kiselgel ≥99.0%
Kiselgel
Purity: 99.5% CAS No.: 7631-86-9 Molecular Formula: SiO2 Formula Weight: 60.09 Physical Form: -325 Mesh Fused Amorphous Powder, S.A. 5m2/g Density: 2.2 pH: 6 Melting Point: 1710° Boiling Point: 2230° Merck Reference: 14,8493 MDL No.: MFCD00011232
Formel: SiO₂
MW: 60,08 g/mol
Kokpunkt: 2230 °C
Smältpunkt: 1710 °C
Densitet: 2,2
Storage Temperature: Omgivande
MDL Number: MFCD00011232
CAS nummer: 7631-86-9
EINECS: 231-545-4
Merck Index: 14,8493