Silicon(II) oxide ≥99.8%, Optical Grade
Leverantör: Thermo Scientific
Varning
Synonyms: Silicon monoxide, Silicon oxide
89430.18 89430.30
89430.18EA 2750 SEK
89430.18 89430.30
Silicon(II) oxide ≥99.8%, Optical Grade
Silicon(II) oxide
Purity: 99.8% CAS No.: 10097-28-6 Molecular Formula: OSi Formula Weight: 44.09 Physical Form: -325 Mesh Powder (typically 10 micron or less) Density: 2.13 Appearance: Black Odour: Odourless Melting Point: 1702° Boiling Point: 1880° Merck Reference: 14,8496 MDL No.: MFCD00151536
Formel: SiO
MW: 44,08 g/mol
Kokpunkt: 1880 °C
Smältpunkt: >1702 °C
Densitet: 2,13
Storage Temperature: Omgivande
MDL Number: MFCD00151536
CAS nummer: 10097-28-6
EINECS: 233-232-8
Merck Index: 14,8496