Bly(II, IV)oxid 98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Minium, Orange lead
221111000 221115000 221110051
ACRO221111000EA 445 SEK
ACRO221111000 ACRO221115000 ACRO221110051
Bly(II, IV)oxid 98%
Bly(II, IV)oxid
Formel: Pb₃O₄
Kokpunkt: 2682 °C (1013 hPa)
Smältpunkt: 500 °C (dec.)
Densitet: 9,1 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00078491
CAS nummer: 1314-41-6
EINECS: 215-235-6
UN: 3288
ADR: 6.1,III
Merck Index: 12,05449
Order Now
Specifikation testresultat
Appearance Orange Powder
Assay ≥97.5 % (PB3O4)
Calcium (Ca) ≤100 ppm
Copper (Cu) ≤50 ppm
Iron (Fe) ≤50 ppm
Magnesium (Mg) ≤100 ppm
Manganese (Mn) ≤10 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤100 ppm
Zinc (Zn) ≤50 ppm