Bly(II)karbonat basic, extra pure
Leverantör: Thermo Scientific
Fara
Synonyms: Lead carbonat basic, White lead, Lead (II) carbonate hydroxide, Lead (II) hydroxide carbonate
222710025 222710050 222715000 222710100
ACRO222710025EA 3575 SEK
ACRO222710025 ACRO222710050 ACRO222715000 ACRO222710100
Bly(II)karbonat basic, extra pure
Bly(II)karbonat basic
Formel: (PbCO₃)₂·Pb(OH)₂
MW: 775,63 g/mol
Smältpunkt: 400 °C (dec.)
Densitet: 6,06 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00078155
CAS nummer: 1319-46-6
EINECS: 215-290-6
UN: 2291
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Titration Complexometric ≥75 % Pb
Insoluble matter ≤0.05 % (in dilute acetic acid)