Bly(II)nitrat ≥99.999%, Puratronic®
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Lead dinitrate
10727.18 10727.30
10727.18EA 3125 SEK
10727.18 10727.30
Bly(II)nitrat ≥99.999%, Puratronic®
Bly(II)nitrat
Purity: 99.999% CAS No.: 10099-74-8 Molecular Formula: Pb(NO3)2 Formula Weight: 331.20 Physical Form: Powder Density: 4.530 Melting Point: 470° dec. Merck Reference: 14,5414 UN No.: 1469 MDL No.: MFCD00011153
Formel: Pb(NO₃)₂
MW: 331.21 g/mol
Smältpunkt: 470 °C (dec.)
Densitet: 4.530
Storage Temperature: Omgivande
MDL Number: MFCD00011153
CAS nummer: 10099-74-8
EINECS: 233-245-9
UN: 1469
ADR: 5.1 II
Merck Index: 14,5414