Bly(II)nitrat 99+%
Leverantör: Thermo Fisher Scientific
Fara
211560010 211565000 211560051
ACRO211560010EA 1775 SEK
ACRO211560010 ACRO211565000 ACRO211560051
Bly(II)nitrat 99+%
Bly(II)nitrat
Formel: Pb(NO₃)₂
Smältpunkt: 470 °C (dec.)
Densitet: 4,53 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00011153
CAS nummer: 10099-74-8
EINECS: 233-245-9
UN: 1469
ADR: 5.1,II
Merck Index: 13,05432
Order Now
Specifikation testresultat
Appearance White Crystalline powder or crystals and/or chunks
Titration Complexometric ≥99.0 %
Chloride (Cl) ≤10 ppm
Iron (Fe) ≤10 ppm